siRNA Details
| Gene Symbol: SLC30A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321036.1 | NP_001307965.1 | 3 | 716 | ||
| NM_013309.5 | NP_037441.2 | 3 | 716 | |||
| XM_011521997.2 | XP_011520299.1 | 3 | 1220 | |||
| XM_017022560.1 | XP_016878049.1 | 2 | 522 | |||
| XM_017022561.1 | XP_016878050.1 | 3 | 714 | |||
| XM_017022562.1 | XP_016878051.1 | 2 | 522 | |||
| GenBank mRNA | AF025409.1 | - | 2 | 404 | ||
| AK290874.1 | - | 2 | 522 | |||
| BC026089.2 | - | 3 | 716 | 4829839 | ||
| BM789500.1 | - | 2 | 286 | |||

SLC30A4
Genomic Map