siRNA Details
| Gene Symbol: RAB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004637.5 | NP_004628.4 | 3 | 310 | ||
| GenBank mRNA | AF498942.1 | - | 2 | 78 | ||
| AK094449.1 | - | 3 | 263 | |||
| AK225691.1 | - | 2 | 57 | |||
| AK290721.1 | - | 3 | 259 | |||
| AK298344.1 | - | 3 | 233 | |||
| AK308934.1 | - | 3 | 253 | |||
| AU122058.1 | - | 3 | 310 | |||
| BC008721.2 | - | 3 | 253 | 2821435 | ||
| BC013728.1 | - | 2 | 93 | 3882701 | ||
| U44104.1 | - | 2 | 78 | |||

RAB7A
Genomic Map