siRNA Details
| Gene Symbol: ST8SIA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005668.5 | NP_005659.1 | 3 | 809 | ||
| NM_175052.2 | NP_778222.1 | 3 | 809 | |||
| XM_005272078.2 | XP_005272135.1 | 3 | 809 | |||
| XM_011543630.1 | XP_011541932.1 | 3 | 809 | |||
| GenBank mRNA | AK292872.1 | - | 3 | 761 | ||
| BC027866.1 | - | 3 | 738 | |||
| BC053657.1 | - | 3 | 790 | 5752337 | ||
| BM921020.1 | - | 3 | 809 | |||
| L41680.1 | - | 3 | 691 | |||

ST8SIA4
Genomic Map