siRNA Details
| Gene Symbol: OR2H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007160.3 | NP_009091.3 | 1 | 714 | ||
| XM_011514912.2 | XP_011513214.1 | 1 | 2096 | |||
| GenBank mRNA | BC069146.1 | - | 1 | 675 | 7217006 | |
| BC101683.1 | - | 1 | 714 | 8069189 | ||
| BC112176.1 | - | 1 | 726 | 8327644 | ||

OR2H2
Genomic Map