siRNA Details
| Gene Symbol: NELFE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002904.5 | NP_002895.3 | 8 | 990 | ||
| XM_006715205.3 | XP_006715268.1 | 8 | 859 | |||
| XM_011514913.2 | XP_011513215.1 | 8 | 930 | |||
| XM_017011299.1 | XP_016866788.1 | 8 | 915 | |||
| GenBank mRNA | AK300717.1 | - | 8 | 773 | ||
| AK300898.1 | - | 8 | 865 | |||
| AK302652.1 | - | 8 | 977 | |||
| AK303152.1 | - | 7 | 1037 | |||
| BC025235.1 | - | 8 | 841 | 4876045 | ||
| BC050617.2 | - | 8 | 857 | 6168937 | ||
| JQ313904.1 | - | 7 | 786 | |||
| L03411.1 | - | 8 | 872 | |||
| X16105.1 | - | 5 | 656 | |||

NELFE
Genomic Map