siRNA Details
| Gene Symbol: TFPI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271003.1 | NP_001257932.1 | 3 | 708 | ||
| NM_001271004.1 | NP_001257933.1 | 3 | 741 | |||
| NM_006528.3 | NP_006519.1 | 3 | 741 | |||
| GenBank mRNA | AK024817.1 | - | 3 | 503 | ||
| AK075425.1 | - | 3 | 502 | |||
| AK075427.1 | - | 3 | 502 | |||
| AK092499.1 | - | 3 | 470 | |||
| AK129833.1 | - | 3 | 503 | |||
| AK313260.1 | - | 3 | 503 | |||
| AY691946.1 | - | 3 | 479 | |||
| BC005330.1 | - | 3 | 503 | 3929933 | ||
| BC034155.1 | - | 2 | 226 | |||
| D29992.1 | - | 3 | 484 | |||
| DA833200.1 | - | 3 | 502 | |||
| L27624.1 | - | 3 | 466 | |||

TFPI2
Genomic Map