siRNA Details
| Gene Symbol: TUSC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006765.3 | NP_006756.2 | 8, 9 | 1286 | ||
| NM_178234.2 | NP_839952.1 | 8, 9 | 1286 | |||
| XM_011544651.2 | XP_011542953.1 | 7, 8 | 829 | |||
| XM_011544652.2 | XP_011542954.1 | 7, 8 | 829 | |||
| XM_017013860.1 | XP_016869349.1 | 7, 8 | 829 | |||
| XM_017013861.1 | XP_016869350.1 | 7, 8 | 829 | |||
| GenBank mRNA | AY562998.1 | - | 1, 2 | 45 | ||
| AY562999.1 | - | 1, 2 | 45 | |||
| BC010370.1 | - | 8, 9 | 1154 | 4334284 | ||
| BT020002.1 | - | 8, 9 | 942 | |||
| U42349.1 | - | 8, 9 | 1099 | |||

TUSC3
Genomic Map