siRNA Details
| Gene Symbol: TUSC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006765.3 | NP_006756.2 | 6 | 1097 | ||
| NM_178234.2 | NP_839952.1 | 6 | 1097 | |||
| XM_011544651.2 | XP_011542953.1 | 5 | 640 | |||
| XM_011544652.2 | XP_011542954.1 | 5 | 640 | |||
| XM_011544654.2 | XP_011542956.1 | 5 | 640 | |||
| XM_017013860.1 | XP_016869349.1 | 5 | 640 | |||
| XM_017013861.1 | XP_016869350.1 | 5 | 640 | |||
| GenBank mRNA | BC010370.1 | - | 6 | 965 | 4334284 | |
| BT020002.1 | - | 6 | 753 | |||
| U42349.1 | - | 6 | 910 | |||

TUSC3
Genomic Map