siRNA Details
| Gene Symbol: NUP214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318324.1 | NP_001305253.1 | 19 | 2746 | ||
| NM_005085.3 | NP_005076.3 | 19 | 2776 | |||
| XM_005272216.3 | XP_005272273.1 | 19 | 2779 | |||
| XM_005272218.3 | XP_005272275.1 | 19 | 2779 | |||
| XM_005272219.3 | XP_005272276.1 | 19 | 2491 | |||
| XM_006717292.3 | XP_006717355.1 | 19 | 2743 | |||
| XM_011519053.2 | XP_011517355.1 | 19 | 2743 | |||
| XM_011519055.2 | XP_011517357.1 | 19 | 2779 | |||
| XM_017015167.1 | XP_016870656.1 | 19 | 2740 | |||
| GenBank mRNA | AB159230.1 | - | 19 | 2635 | ||
| AK310406.1 | - | 7 | 821 | |||
| AL832609.2 | - | 9 | 1415 | |||
| BC045620.1 | - | 19 | 2720 | 5268515 | ||
| BC105998.1 | - | 19 | 2721 | 30531438 | ||
| BX537986.1 | - | 9 | 1166 | |||
| D14689.1 | - | 19 | 2654 | |||
| X64228.1 | - | 19 | 2726 | |||

NUP214
Genomic Map