siRNA Details
| Gene Symbol: CCDC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005436.4 | NP_005427.2 | 9 | 1486 | ||
| GenBank mRNA | AA112388.1 | - | 2 | 131 | ||
| AK056848.1 | - | 3 | 1306 | |||
| AK292593.1 | - | 9 | 1486 | |||
| BC036757.2 | - | 9 | 1325 | 5549623 | ||
| BC064391.1 | - | 9 | 1312 | 5745698 | ||
| S72869.1 | - | 9 | 1290 | |||

CCDC6
Genomic Map