siRNA Details
| Gene Symbol: SHOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001269039.2 | NP_001255968.1 | 2 | 874 | ||
| NM_001324336.1 | NP_001311265.1 | 2 | 831 | |||
| NM_001324337.1 | NP_001311266.1 | 3 | 1059 | |||
| NM_007373.3 | NP_031399.2 | 2 | 874 | |||
| XM_017016702.1 | XP_016872191.1 | 2 | 779 | |||
| XM_017016703.1 | XP_016872192.1 | 2 | 6596 | |||
| XM_017016704.1 | XP_016872193.1 | 2 | 778 | |||
| GenBank mRNA | AB020669.1 | - | 2 | 802 | ||
| AF054828.1 | - | 2 | 796 | |||
| AF068920.1 | - | 1 | 525 | |||
| AK090820.1 | - | 1 | 33 | |||
| AK292833.1 | - | 3 | 1000 | |||
| BC044752.1 | - | 2 | 810 | |||
| BC050445.1 | - | 1 | 729 | 6067669 | ||
| BX649055.1 | - | 3 | 642 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001168505, NM_019658, BC026364, AF068921, AK146447, and others.

SHOC2
Genomic Map