siRNA Details
| Gene Symbol: COIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004645.2 | NP_004636.1 | 2 | 839 | ||
| GenBank mRNA | AK313616.1 | - | 2 | 821 | ||
| BC010385.1 | - | 2 | 836 | 4275993 | ||
| M58411.1 | - | 1 | 296 | |||
| U06632.1 | - | 2 | 830 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

COIL| Gene Symbol: COIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004645.2 | NP_004636.1 | 2 | 839 | ||
| GenBank mRNA | AK313616.1 | - | 2 | 821 | ||
| BC010385.1 | - | 2 | 836 | 4275993 | ||
| M58411.1 | - | 1 | 296 | |||
| U06632.1 | - | 2 | 830 | |||