siRNA Details
| Gene Symbol: SF3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007165.4 | NP_009096.2 | 5, 6 | 477 | ||
| GenBank mRNA | AK314815.1 | - | 5, 6 | 470 | ||
| BC004434.1 | - | 5, 6 | 447 | 2820900 | ||
| BC009903.2 | - | 5, 6 | 470 | 2959124 | ||
| BC015804.1 | - | 5, 6 | 430 | |||

SF3A2
Genomic Map