siRNA Details
| Gene Symbol: RBM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204466.1 | NP_001191395.1 | 10 | 1303 | ||
| NM_001204467.1 | NP_001191396.1 | 11 | 1531 | |||
| NM_001204468.1 | NP_001191397.1 | 11 | 1565 | |||
| NM_005676.4 | NP_005667.2 | 11 | 1534 | |||
| NM_152856.2 | NP_690595.1 | 10 | 1300 | |||
| XM_005272677.4 | XP_005272734.1 | 11 | 1584 | |||
| XM_005272678.4 | XP_005272735.1 | 10 | 1118 | |||
| XM_005272679.4 | XP_005272736.1 | 10 | 1115 | |||
| XM_006724563.1 | XP_006724626.1 | 7 | 751 | |||
| XM_011543989.2 | XP_011542291.1 | 3 | 475 | |||
| XM_017029884.1 | XP_016885373.1 | 10 | 1454 | |||
| XM_017029885.1 | XP_016885374.1 | 10 | 1226 | |||
| GenBank mRNA | AK292758.1 | - | 10 | 1254 | ||
| AL137421.1 | - | 12 | 1423 | |||
| BC000681.2 | - | 8 | 756 | |||
| BC003089.1 | - | 10 | 1239 | 3536058 | ||
| BC004181.2 | - | 11 | 1487 | 2961094 | ||
| BC008733.2 | - | 11 | 1519 | 3163064 | ||
| BC024153.2 | - | 10 | 1149 | 3845258 | ||
| BX537969.1 | - | 11 | 1516 | |||
| D50912.1 | - | 11 | 1382 | |||
| KU178411.1 | - | 9 | 907 | |||
| KU178412.1 | - | 10 | 1141 | |||

RBM10
Genomic Map