siRNA Details
| Gene Symbol: RBM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204466.1 | NP_001191395.1 | 3 | 559 | ||
| NM_001204467.1 | NP_001191396.1 | 3 | 559 | |||
| NM_001204468.1 | NP_001191397.1 | 3 | 590 | |||
| NM_005676.4 | NP_005667.2 | 3 | 559 | |||
| NM_152856.2 | NP_690595.1 | 3 | 559 | |||
| XM_005272677.4 | XP_005272734.1 | 3 | 612 | |||
| XM_005272678.4 | XP_005272735.1 | 3 | 374 | |||
| XM_005272679.4 | XP_005272736.1 | 3 | 374 | |||
| XM_006724563.1 | XP_006724626.1 | 1 | 81 | |||
| XM_017029884.1 | XP_016885373.1 | 3 | 553 | |||
| XM_017029885.1 | XP_016885374.1 | 3 | 328 | |||
| GenBank mRNA | AK000962.1 | - | 3 | 527 | ||
| AK098115.1 | - | 81 | ||||
| AK225175.1 | - | 3 | 518 | |||
| AK292758.1 | - | 3 | 510 | |||
| AL137421.1 | - | 4 | 448 | |||
| BC003089.1 | - | 3 | 498 | 3536058 | ||
| BC004181.2 | - | 3 | 512 | 2961094 | ||
| BC008733.2 | - | 3 | 544 | 3163064 | ||
| BC024153.2 | - | 2 | 174 | 3845258 | ||
| BX537969.1 | - | 3 | 541 | |||
| D50912.1 | - | 3 | 410 | |||
| KU178411.1 | - | 2 | 166 | |||
| KU178412.1 | - | 2 | 166 | |||

RBM10
Genomic Map