siRNA Details
Gene Symbol: TMEM187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003492.2 | NP_003483.1 | 2 | 630 | ||
XM_011531199.2 | XP_011529501.1 | 2 | 645 | |||
GenBank mRNA | AK314938.1 | - | 2 | 410 | ||
BC008203.1 | - | 2 | 378 | 3868838 | ||
BC017026.1 | - | 2 | 365 | 3847234 | ||
DQ655925.2 | - | 3 | 363 | |||
X92475.1 | - | 2 | 380 |
