siRNA Details
| Gene Symbol: SLC10A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142391.2 | NP_001135863.1 | 4 | 1064 | ||
| NM_001142392.2 | NP_001135864.1 | 3 | 1151 | |||
| NM_019848.4 | NP_062822.1 | 2 | 1386 | |||
| XM_005277913.4 | XP_005277970.1 | 2 | 1808 | |||
| XM_006724847.3 | XP_006724910.1 | 2 | 1329 | |||
| XM_006724848.3 | XP_006724911.1 | 3 | 1242 | |||
| XM_011531200.2 | XP_011529502.2 | 2 | 1361 | |||
| XM_011531201.2 | XP_011529503.1 | 2 | 1065 | |||
| GenBank mRNA | BC004966.1 | - | 4 | 988 | 3503242 | |

SLC10A3
Genomic Map