siRNA Details
| Gene Symbol: CDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134419.1 | NP_001127891.1 | 5 | 543 | ||
| NM_001134420.1 | NP_001127892.1 | 5 | 671 | |||
| NM_003503.3 | NP_003494.1 | 5 | 577 | |||
| XM_005271241.2 | XP_005271298.1 | 5 | 539 | |||
| XM_005271244.2 | XP_005271301.1 | 5 | 577 | |||
| XM_017002425.1 | XP_016857914.1 | 5 | 539 | |||
| XM_017002426.1 | XP_016857915.1 | 5 | 522 | |||
| XM_017002427.1 | XP_016857916.1 | 4 | 385 | |||
| GenBank mRNA | AB003698.1 | - | 5 | 544 | ||
| AB209337.1 | - | 4 | 1253 | |||
| AF005209.1 | - | 4 | 412 | |||
| AF015592.1 | - | 5 | 557 | |||
| AK298963.1 | - | 4 | 489 | |||
| AK312726.1 | - | 5 | 633 | |||
| BC110526.1 | - | 5 | 500 | 40034178 | ||
| BC110527.1 | - | 5 | 534 | 40034179 | ||
| BC111044.1 | - | 5 | 557 | 5518616 | ||

CDC7
Genomic Map