siRNA Details
| Gene Symbol: HIST1H4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003539.3 | NP_003530.1 | 1 | 24 | ||
| GenBank mRNA | BC128104.1 | - | 1 | 24 | 40112084 | |
| BC128105.1 | - | 1 | 23 | |||
| BC143045.1 | - | 1 | 24 | 40112087 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H4D| Gene Symbol: HIST1H4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003539.3 | NP_003530.1 | 1 | 24 | ||
| GenBank mRNA | BC128104.1 | - | 1 | 24 | 40112084 | |
| BC128105.1 | - | 1 | 23 | |||
| BC143045.1 | - | 1 | 24 | 40112087 | ||