siRNA Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001326578.1 | NP_001313507.1 | 10 | 1079 | ||
| NM_001326579.1 | NP_001313508.1 | 10 | 866 | |||
| NM_003569.2 | NP_003560.2 | 10 | 1002 | |||
| NR_137169.1 | 9 | 929 | ||||
| GenBank mRNA | AK294911.1 | - | 10 | 751 | ||
| AK301464.1 | - | 4 | 365 | |||
| BC011975.1 | - | 10 | 866 | 3860059 | ||
| U77942.1 | - | 10 | 831 | |||

STX7
Genomic Map