siRNA Details
| Gene Symbol: ZNF282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303481.1 | NP_001290410.1 | 2 | 356 | ||
| NM_003575.3 | NP_003566.1 | 2 | 356 | |||
| XM_006716151.3 | XP_006716214.1 | 2 | 350 | |||
| GenBank mRNA | AK299280.1 | - | 2 | 268 | ||
| AK316282.1 | - | 2 | 243 | |||
| AY927505.1 | - | 1 | 29 | |||
| BC041704.1 | - | 2 | 83 | |||
| BC055287.1 | - | 2 | 146 | |||
| BC073805.1 | - | 2 | 249 | 6645861 | ||
| CV573257.1 | - | 2 | 333 | |||
| D30612.1 | - | 2 | 369 | |||

ZNF282
Genomic Map