siRNA Details
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 7, 8 | 889 | ||
NM_001278513.1 | NP_001265442.1 | 7, 8 | 1033 | |||
NM_001278514.1 | NP_001265443.1 | 7, 8 | 701 | |||
NM_003589.2 | NP_003580.1 | 7, 8 | 715 | |||
XM_011537523.2 | XP_011535825.1 | 6, 7 | 1249 | |||
GenBank mRNA | AB012193.1 | - | 1, 2 | 63 | ||
AB178950.1 | - | 7, 8 | 766 | |||
AF077188.1 | - | 7, 8 | 626 | |||
AK314835.1 | - | 7, 8 | 988 | |||
AL833355.1 | - | 6, 7 | 653 | |||
AY365124.1 | - | 7, 8 | 766 | |||
BC008308.1 | - | 7, 8 | 777 | 3537176 |
