siRNA Details
| Gene Symbol: TPST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003596.3 | NP_003587.1 | 2 | 772 | ||
| XM_005250642.2 | XP_005250699.1 | 2 | 768 | |||
| XM_011516634.2 | XP_011514936.1 | 2 | 768 | |||
| XM_011516635.2 | XP_011514937.1 | 2 | 768 | |||
| XM_017012724.1 | XP_016868213.1 | 3 | 855 | |||
| XM_017012725.1 | XP_016868214.1 | 3 | 855 | |||
| XM_017012726.1 | XP_016868215.1 | 2 | 768 | |||
| XM_017012727.1 | XP_016868216.1 | 2 | 768 | |||
| GenBank mRNA | AF038009.1 | - | 1 | 501 | ||
| AK313098.1 | - | 2 | 710 | |||
| BC013188.1 | - | 2 | 747 | 3918922 | ||
| CR542060.1 | - | 1 | 420 | |||
| CR542080.1 | - | 1 | 420 | |||

TPST1
Genomic Map