siRNA Details
| Gene Symbol: MAP4K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270425.1 | NP_001257354.1 | 29 | 2586 | ||
| NM_003618.3 | NP_003609.2 | 30 | 2649 | |||
| GenBank mRNA | AK021553.1 | - | 2 | 153 | ||
| AK291467.1 | - | 30 | 2615 | |||
| AK299889.1 | - | 22 | 1678 | |||
| AK314487.1 | - | 29 | 2423 | |||
| AK316144.1 | - | 3 | 229 | |||
| BC071579.1 | - | 29 | 2578 | 5296205 | ||

MAP4K3
Genomic Map