siRNA Details
| Gene Symbol: PEX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003630.2 | NP_003621.1 | 5 | 689 | ||
| GenBank mRNA | AB035307.1 | - | 5 | 573 | ||
| AJ001625.1 | - | 5 | 490 | |||
| AJ131389.1 | - | 5 | 486 | |||
| AK025644.1 | - | 555 | ||||
| AK300907.1 | - | 5 | 555 | |||
| AY277600.1 | - | 5 | 446 | |||
| BC014551.1 | - | 5 | 627 | 4333011 | ||
| BC015506.1 | - | 5 | 588 | 3914956 | ||
| CR542062.1 | - | 5 | 427 | |||

PEX3
Genomic Map