siRNA Details
| Gene Symbol: CPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014447.2 | NP_001014447.1 | 7 | 1262 | ||
| NM_001014448.2 | NP_001014448.1 | 7 | 1525 | |||
| NM_003652.3 | NP_003643.2 | 6 | 1229 | |||
| GenBank mRNA | AK056317.1 | - | 7 | 1261 | ||
| AK127844.1 | - | 4 | 2598 | |||
| BC006393.2 | - | 7 | 1429 | |||
| BC018913.2 | - | 7 | 1429 | |||
| BC080539.1 | - | 7 | 1196 | 5020194 | ||
| U83411.1 | - | 6 | 1094 | |||

CPZ
Genomic Map