siRNA Details
| Gene Symbol: CPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014447.2 | NP_001014447.1 | 3 | 415 | ||
| NM_001014448.2 | NP_001014448.1 | 3 | 678 | |||
| NM_003652.3 | NP_003643.2 | 2 | 382 | |||
| GenBank mRNA | AK056317.1 | - | 3 | 415 | ||
| BC006393.2 | - | 3 | 582 | |||
| BC018913.2 | - | 3 | 582 | |||
| BC080539.1 | - | 3 | 349 | 5020194 | ||
| BX338482.2 | - | 2 | 235 | |||
| DA050438.1 | - | 3 | 540 | |||
| DA704214.1 | - | 3 | 415 | |||
| U83411.1 | - | 2 | 247 | |||

CPZ
Genomic Map