siRNA Details
| Gene Symbol: CPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014447.2 | NP_001014447.1 | 3 | 609 | ||
| NM_001014448.2 | NP_001014448.1 | 3 | 872 | |||
| NM_003652.3 | NP_003643.2 | 2 | 576 | |||
| GenBank mRNA | AK056317.1 | - | 3 | 609 | ||
| BC006393.2 | - | 3 | 776 | |||
| BC018913.2 | - | 3 | 776 | |||
| BC080539.1 | - | 3 | 543 | 5020194 | ||
| BX338482.2 | - | 2 | 429 | |||
| CV026874.1 | - | 1 | 24 | |||
| U83411.1 | - | 2 | 441 | |||

CPZ
Genomic Map