siRNA Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003695.2 | NP_003686.1 | 3 | 257 | ||
| GenBank mRNA | AK312164.1 | - | 3 | 204 | ||
| BC022806.1 | - | 3 | 184 | |||
| BC031330.1 | - | 3 | 257 | 5428818 | ||
| BC034542.1 | - | 1 | 878 | |||
| BM019592.1 | - | 3 | 268 | |||
| BU689475.1 | - | 1 | 558 | |||
| X82693.1 | - | 3 | 192 | |||

LY6D
Genomic Map