siRNA Details
| Gene Symbol: NOP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291978.1 | NP_001278907.1 | 10 | 1661 | ||
| NM_001291979.1 | NP_001278908.1 | 10 | 1661 | |||
| NM_003703.2 | NP_003694.1 | 10 | 1661 | |||
| GenBank mRNA | AB000467.1 | - | 8 | 1057 | ||
| AF040965.1 | - | 10 | 1523 | |||
| AK025692.1 | - | 4 | 526 | |||
| AK292939.1 | - | 10 | 1661 | |||
| BC009760.2 | - | 6 | 855 | |||
| BC026035.1 | - | 10 | 1488 | 4878317 | ||
| BC053635.1 | - | 10 | 1540 | 5114280 | ||

NOP14
Genomic Map