siRNA Details
| Gene Symbol: NOP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291978.1 | NP_001278907.1 | 17 | 2576 | ||
| NM_003703.2 | NP_003694.1 | 17 | 2576 | |||
| GenBank mRNA | AB000467.1 | - | 15 | 1972 | ||
| AF040965.1 | - | 17 | 2438 | |||
| AK024755.1 | - | 7 | 885 | |||
| AK025692.1 | - | 11 | 1441 | |||
| AK292939.1 | - | 17 | 2576 | |||
| BC053635.1 | - | 17 | 2455 | 5114280 | ||

NOP14
Genomic Map