siRNA Details
| Gene Symbol: CDK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003718.4 | NP_003709.3 | 2 | 2128 | ||
| NM_031267.3 | NP_112557.2 | 2 | 2128 | |||
| XM_011515597.2 | XP_011513899.1 | 2 | 2987 | |||
| XM_017012750.1 | XP_016868239.1 | 2 | 2987 | |||
| XM_017012751.1 | XP_016868240.1 | 2 | 2987 | |||
| GenBank mRNA | AJ297709.1 | - | 2 | 1978 | ||
| AJ297710.1 | - | 2 | 1978 | |||
| AK092534.1 | - | 2 | 2128 | |||
| BC001274.1 | - | 1 | 398 | |||
| BC080601.1 | - | 2 | 2451 | |||
| M80629.1 | - | 2 | 775 | |||

CDK13
Genomic Map