siRNA Details
| Gene Symbol: CDK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003718.4 | NP_003709.3 | 11 | 3197 | ||
| NM_031267.3 | NP_112557.2 | 11 | 3197 | |||
| XM_017012750.1 | XP_016868239.1 | 12 | 4146 | |||
| XM_017012751.1 | XP_016868240.1 | 12 | 4146 | |||
| GenBank mRNA | AB058694.1 | - | 5 | 2613 | ||
| AJ297709.1 | - | 11 | 3047 | |||
| AJ297710.1 | - | 11 | 3047 | |||
| AK223053.1 | - | 5 | 350 | |||
| BF707421.1 | - | 4 | 244 | |||
| M80629.1 | - | 11 | 1844 | |||

CDK13
Genomic Map