siRNA Details
| Gene Symbol: UNC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003728.3 | NP_003719.3 | 3, 4 | 834 | ||
| XM_005263321.3 | XP_005263378.1 | 3, 4 | 834 | |||
| XM_017008789.1 | XP_016864278.1 | 4, 5 | 7251 | |||
| XM_017008790.1 | XP_016864279.1 | 3, 4 | 479 | |||
| GenBank mRNA | AF055634.1 | - | 3, 4 | 635 | ||
| AK290500.1 | - | 3, 4 | 645 | |||
| AK308421.1 | - | 3, 4 | 597 | |||
| BC041156.1 | - | 3, 4 | 830 | 5208108 | ||

UNC5C
Genomic Map