siRNA Details
| Gene Symbol: RTCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130841.1 | NP_001124313.1 | 4 | 474 | ||
| NM_003729.3 | NP_003720.1 | 3 | 435 | |||
| XM_005271298.4 | XP_005271355.1 | 3 | 432 | |||
| XM_005271299.4 | XP_005271356.1 | 3 | 421 | |||
| XM_017002669.1 | XP_016858158.1 | 3 | 178 | |||
| GenBank mRNA | AK292366.1 | - | 3 | 384 | ||
| AK299793.1 | - | 3 | 436 | |||
| AK307445.1 | - | 3 | 356 | |||
| AU310497.1 | - | 3 | 436 | |||
| BC012604.1 | - | 4 | 388 | 4281547 | ||
| DA361597.1 | - | 3 | 377 | |||
| Y11651.1 | - | 3 | 387 | |||

RTCA
Genomic Map