siRNA Details
| Gene Symbol: KCNK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003740.3 | NP_003731.1 | 3 | 795 | ||
| XM_005249456.1 | XP_005249513.1 | 3 | 528 | |||
| XM_006715235.2 | XP_006715298.1 | 1 | 69 | |||
| GenBank mRNA | AF084830.1 | - | 3 | 770 | ||
| AK314298.1 | - | 3 | 528 | |||
| BC060793.1 | - | 3 | 795 | 30348251 | ||
| BC069573.1 | - | 3 | 502 | 7262361 | ||
| EU978936.1 | - | 3 | 581 | |||

KCNK5
Genomic Map