siRNA Details
| Gene Symbol: RIPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317061.1 | NP_001303990.1 | 7 | 1106 | ||
| NM_003804.4 | NP_003795.2 | 5 | 828 | |||
| XM_005249457.3 | XP_005249514.2 | 5 | 751 | |||
| XM_006715237.3 | XP_006715300.2 | 5 | 751 | |||
| XM_017011403.1 | XP_016866892.1 | 2 | 860 | |||
| XM_017011405.1 | XP_016866894.1 | 5 | 749 | |||
| GenBank mRNA | AB208926.1 | - | 7 | 1015 | ||
| AK304701.1 | - | 3 | 417 | |||
| AK314176.1 | - | 5 | 828 | |||
| BC126254.1 | - | 4 | 622 | 8991970 | ||
| BC126256.1 | - | 4 | 622 | 8991972 | ||
| U50062.1 | - | 4 | 596 | |||

RIPK1
Genomic Map