siRNA Details
| Gene Symbol: MTMR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306144.1 | NP_001293073.1 | 7 | 737 | ||
| NM_001306145.1 | NP_001293074.1 | 6 | 713 | |||
| NM_003828.3 | NP_003819.1 | 6 | 713 | |||
| XM_005274765.3 | XP_005274822.1 | 9 | 777 | |||
| XM_005274766.3 | XP_005274823.1 | 8 | 725 | |||
| XM_005274767.3 | XP_005274824.1 | 7 | 744 | |||
| XM_006724855.3 | XP_006724918.1 | 6 | 713 | |||
| XM_011531209.2 | XP_011529511.1 | 6 | 473 | |||
| XM_011531210.2 | XP_011529512.1 | 9 | 779 | |||
| XM_017029920.1 | XP_016885409.1 | 9 | 777 | |||
| XM_017029921.1 | XP_016885410.1 | 7 | 508 | |||
| XM_017029922.1 | XP_016885411.1 | 7 | 520 | |||
| XM_017029923.1 | XP_016885412.1 | 4 | 300 | |||
| XM_017029924.1 | XP_016885413.1 | 7 | 744 | |||
| GenBank mRNA | AF057354.1 | - | 6 | 537 | ||
| AJ224979.1 | - | 6 | 539 | |||
| AK001816.1 | - | 4 | 242 | |||
| AK226109.1 | - | 7 | 691 | |||
| AK295738.1 | - | 7 | 532 | |||
| BC032947.1 | - | 6 | 710 | 4837744 | ||
| U58032.1 | - | 3 | 318 | |||

MTMR1
Genomic Map