siRNA Details
| Gene Symbol: RGS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286485.1 | NP_001273414.1 | 13 | 1011 | ||
| NM_001286486.1 | NP_001273415.1 | 12 | 879 | |||
| NM_003834.2 | NP_003825.1 | 14 | 1055 | |||
| NM_183337.2 | NP_899180.1 | 14 | 1044 | |||
| XM_011522719.2 | XP_011521021.1 | 15 | 1138 | |||
| XM_011522720.2 | XP_011521022.1 | 15 | 1078 | |||
| GenBank mRNA | AB016929.1 | - | 13 | 979 | ||
| AB209582.1 | - | 6 | 2952 | |||
| AF035153.1 | - | 14 | 1015 | |||
| AF035154.1 | - | 14 | 1026 | |||
| AK091701.1 | - | 12 | 876 | |||
| BC141803.1 | - | 14 | 1019 | 40147243 | ||

RGS11
Genomic Map