siRNA Details
Gene Symbol: FGF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003868.2 | NP_003859.1 | 3 | 609 | ||
GenBank mRNA | AB009391.1 | - | 3 | 609 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FGF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003868.2 | NP_003859.1 | 3 | 609 | ||
GenBank mRNA | AB009391.1 | - | 3 | 609 |