siRNA Details
Gene Symbol: FCGBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003890.2 | NP_003881.2 | 6 | 2937 | ||
GenBank mRNA | D84239.1 | - | 2937 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FCGBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003890.2 | NP_003881.2 | 6 | 2937 | ||
GenBank mRNA | D84239.1 | - | 2937 |