siRNA Details
| Gene Symbol: SYNJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160302.1 | NP_001153774.1 | 32 | 4656 | ||
| NM_001160306.1 | NP_001153778.1 | 28 | 4539 | |||
| NM_003895.3 | NP_003886.3 | 32 | 4672 | |||
| NM_203446.2 | NP_982271.2 | 33 | 4696 | |||
| XM_017028494.1 | XP_016883983.1 | 31 | 4614 | |||
| XM_017028495.1 | XP_016883984.1 | 31 | 4672 | |||
| XM_017028496.1 | XP_016883985.1 | 30 | 4575 | |||
| XM_017028497.1 | XP_016883986.1 | 31 | 4712 | |||
| XM_017028498.1 | XP_016883987.1 | 29 | 4536 | |||
| XM_017028499.1 | XP_016883988.1 | 28 | 4546 | |||
| XM_017028500.1 | XP_016883989.1 | 33 | 4686 | |||
| XM_017028501.1 | XP_016883990.1 | 33 | 4683 | |||
| XM_017028502.1 | XP_016883991.1 | 32 | 4635 | |||
| XM_017028503.1 | XP_016883992.1 | 31 | 4599 | |||
| XM_017028504.1 | XP_016883993.1 | 30 | 4560 | |||
| XM_017028505.1 | XP_016883994.1 | 29 | 4570 | |||
| GenBank mRNA | AB020717.2 | - | 31 | 4598 | ||
| AF009039.1 | - | 33 | 4628 | |||
| AF009040.1 | - | 32 | 4604 | |||
| AK304044.1 | - | 7 | 1519 | |||
| BC098395.1 | - | 28 | 4506 | |||
| BC136603.1 | - | 28 | 4465 | 9020593 | ||
| DQ421853.1 | - | 6 | 1371 | |||

SYNJ1
Genomic Map