siRNA Details
| Gene Symbol: CDC123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D14878.1 | - | 12 | 1240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC123| Gene Symbol: CDC123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D14878.1 | - | 12 | 1240 | ||