siRNA Details
| Gene Symbol: CDC123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006023.2 | NP_006014.2 | 6, 7 | 732 | ||
| XM_005252638.3 | XP_005252695.1 | 5, 6 | 453 | |||
| GenBank mRNA | AK225634.1 | - | 6, 7 | 698 | ||
| AK289362.1 | - | 6, 7 | 469 | |||
| BC001600.1 | - | 6, 7 | 482 | 3542611 | ||
| BC009598.1 | - | 6, 7 | 497 | 4134376 | ||
| CR456966.1 | - | 6, 7 | 448 | |||
| D14878.1 | - | 6, 7 | 728 | |||
| U27112.1 | - | 6, 7 | 510 | |||

CDC123
Genomic Map