siRNA Details
| Gene Symbol: RAB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135662.1 | NP_001129134.1 | 6 | 812 | ||
| NM_001135663.1 | NP_001129135.1 | 4 | 1027 | |||
| NM_001135664.1 | NP_001129136.1 | 5 | 659 | |||
| NM_003929.2 | NP_003920.1 | 6 | 913 | |||
| XM_005245569.1 | XP_005245626.1 | 6 | 811 | |||
| XM_005245570.1 | XP_005245627.1 | 6 | 864 | |||
| XM_005245571.1 | XP_005245628.1 | 6 | 789 | |||
| XM_006711605.3 | XP_006711668.1 | 5 | 555 | |||
| XM_006711606.2 | XP_006711669.1 | 5 | 519 | |||
| XM_017002748.1 | XP_016858237.1 | 5 | 814 | |||
| XM_017002749.1 | XP_016858238.1 | 5 | 792 | |||
| XM_017002750.1 | XP_016858239.1 | 5 | 724 | |||
| GenBank mRNA | AK223514.1 | - | 5 | 689 | ||
| AK303879.1 | - | 4 | 778 | |||
| AK308359.1 | - | 5 | 628 | |||
| AK311085.1 | - | 4 | 536 | |||
| AK313000.1 | - | 6 | 801 | |||
| BC002585.2 | - | 6 | 746 | 3140744 | ||
| BT019540.1 | - | 5 | 573 | |||
| BX647960.1 | - | 6 | 878 | |||
| CR542010.1 | - | 5 | 573 | |||
| D84488.1 | - | 5 | 613 | |||

RAB29
Genomic Map