siRNA Details
| Gene Symbol: HSPB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK314215.1 | - | 1 | 466 | ||
| BC126272.1 | - | 1 | 320 | 8991988 | ||
| BC132869.1 | - | 1 | 332 | 40146891 | ||
| U15590.1 | - | 1 | 451 | |||
| Y17782.1 | - | 1 | 424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSPB3| Gene Symbol: HSPB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK314215.1 | - | 1 | 466 | ||
| BC126272.1 | - | 1 | 320 | 8991988 | ||
| BC132869.1 | - | 1 | 332 | 40146891 | ||
| U15590.1 | - | 1 | 451 | |||
| Y17782.1 | - | 1 | 424 | |||