siRNA Details
| Gene Symbol: CH25H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003956.3 | NP_003947.1 | 1 | 241 | ||
| GenBank mRNA | AF059214.1 | - | 1 | 229 | ||
| AK314865.1 | - | 1 | 246 | |||
| BC017843.1 | - | 1 | 241 | 4619357 | ||
| BC072430.1 | - | 1 | 221 | 5589142 | ||

CH25H
Genomic Map