siRNA Details
| Gene Symbol: UBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003968.3 | NP_003959.3 | 10 | 766 | ||
| NM_198195.1 | NP_937838.1 | 9 | 724 | |||
| XM_011534210.1 | XP_011532512.1 | 9 | 680 | |||
| XM_011534211.1 | XP_011532513.1 | 8 | 633 | |||
| GenBank mRNA | AB012190.1 | - | 8 | 683 | ||
| AF046024.1 | - | 9 | 767 | |||
| AK002159.1 | - | 9 | 711 | |||
| AK289392.1 | - | 10 | 752 | |||
| AK298878.1 | - | 5 | 439 | |||
| AK314751.1 | - | 10 | 749 | |||
| AK316540.1 | - | 7 | 603 | |||
| AL117566.1 | - | 10 | 762 | |||
| BC022853.1 | - | 10 | 765 | 4073511 | ||
| CR533537.1 | - | 10 | 746 | |||

UBA3
Genomic Map