siRNA Details
| Gene Symbol: SH2D2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161441.1 | NP_001154913.1 | 7 | 948 | ||
| NM_001161442.1 | NP_001154914.1 | 7 | 887 | |||
| NM_001161443.1 | NP_001154915.1 | 6 | 797 | |||
| NM_001161444.1 | NP_001154916.1 | 7 | 918 | |||
| NM_003975.3 | NP_003966.2 | 7 | 918 | |||
| XM_006711615.2 | XP_006711678.1 | 7 | 936 | |||
| XM_017002762.1 | XP_016858251.1 | 7 | 965 | |||
| XM_017002763.1 | XP_016858252.1 | 7 | 956 | |||
| XM_017002764.1 | XP_016858253.1 | 7 | 903 | |||
| XM_017002765.1 | XP_016858254.1 | 7 | 916 | |||
| XM_017002766.1 | XP_016858255.1 | 7 | 906 | |||
| GenBank mRNA | AF051325.1 | - | 7 | 840 | ||
| AF097744.1 | - | 7 | 817 | |||
| AJ000553.1 | - | 7 | 864 | |||
| AK222737.1 | - | 7 | 841 | |||
| AK313706.1 | - | 7 | 912 | |||
| AY763098.1 | - | 6 | 797 | |||
| AY763099.1 | - | 7 | 894 | |||
| AY763100.1 | - | 7 | 824 | |||
| BC012107.1 | - | 7 | 880 | 4636736 | ||

SH2D2A
Genomic Map