siRNA Details
| Gene Symbol: CLDN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185072.2 | NP_001172001.1 | 4 | 941 | ||
| NM_001185073.2 | NP_001172002.1 | 3 | 902 | |||
| NM_012129.4 | NP_036261.1 | 3 | 898 | |||
| GenBank mRNA | AK312480.1 | - | 4 | 826 | ||
| AL136770.1 | - | 3 | 750 | |||
| BC036754.2 | - | 2 | 687 | 5201423 | ||
| BC068532.1 | - | 3 | 784 | 5294796 | ||
| CR533470.1 | - | 1 | 559 | |||

CLDN12
Genomic Map